Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243293.1 | NP_001230222.1 | 4 - 5 | 736 | 86 | |
GenBank mRNA | AK348219.1 | - | 4 - 5 | 736 | 86 | |
AK391940.1 | - | 4 - 5 | 721 | 86 | ||
AY610027.1 | - | 4 - 5 | 681 | 86 | ||
FS700419.1 | - | 4 - 5 | 702 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |