Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243338.1 | NP_001230267.1 | 3 - 4 | 551 | 83 | |
XM_005671055.3 | XP_005671112.1 | 610 | 83 | |||
GenBank mRNA | AK346162.1 | - | 3 - 4 | 612 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |