Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243341.1 | NP_001230270.1 | 1 - 2 | 206 | 111 | |
XM_021068495.1 | XP_020924154.1 | 1501 | 111 | |||
GenBank mRNA | AK230916.1 | - | 1 - 2 | 111 | ||
BP463051.1 | - | 1 - 2 | 208 | 111 | ||
BW980083.1 | - | 1 - 2 | 167 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |