Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243357.1 | NP_001230286.1 | 1 - 2 | 114 | 87 | |
GenBank mRNA | EW506027.2 | - | 1 - 2 | 117 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |