Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243357.1 | NP_001230286.1 | 5 - 6 | 861 | 63 | |
XM_013990046.2 | XP_013845500.1 | 1356 | 63 | |||
XM_013990047.2 | XP_013845501.1 | 1207 | 63 | |||
GenBank mRNA | AK351275.1 | - | 4 - 5 | 1133 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |