Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243358.1 | NP_001230287.1 | 3 - 4 | 594 | 114 | |
GenBank mRNA | AK231277.1 | - | 3 - 4 | 574 | 114 | |
CB479849.1 | - | 4 - 5 | 535 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |