Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243360.1 | NP_001230289.1 | 1 - 2 | 111 | 98 | |
GenBank mRNA | AK350235.1 | - | 1 - 2 | 75 | 98 | |
BP144127.1 | - | 1 - 2 | 111 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |