Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243360.1 | NP_001230289.1 | 3 - 4 | 200 | 79 | |
GenBank mRNA | AK350235.1 | - | 3 - 4 | 164 | 79 | |
BP144127.1 | - | 3 - 4 | 200 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |