Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243362.1 | NP_001230291.1 | 3 - 4 | 640 | 121 | |
XM_021086851.1 | XP_020942510.1 | 745 | 121 | |||
GenBank mRNA | CK462109.1 | - | 3 - 4 | 296 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |