Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243376.1 | NP_001230305.1 | 2 - 3 | 363 | 90 | |
XM_005659948.3 | XP_005660005.1 | 307 | 90 | |||
XM_013993391.2 | XP_013848845.1 | 408 | 90 | |||
GenBank mRNA | AK343636.1 | - | 2 - 3 | 259 | 90 | |
AY609449.1 | - | 2 - 3 | 217 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |