Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243397.1 | NP_001230326.1 | 3 - 4 | 369 | 78 | |
XM_005652508.3 | XP_005652565.1 | 438 | 78 | |||
GenBank mRNA | AK344907.1 | - | 3 - 4 | 372 | 78 | |
BP441548.1 | - | 3 - 4 | 369 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |