Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243412.1 | NP_001230341.1 | 2 - 3 | 195 | 116 | |
XM_013988430.2 | XP_013843884.1 | 920 | 116 | |||
XM_013988431.2 | XP_013843885.1 | 94 | 116 | |||
XM_021090922.1 | XP_020946581.1 | 920 | 116 | |||
GenBank mRNA | AK343836.1 | - | 2 - 3 | 179 | 116 | |
AK400126.1 | - | 2 - 3 | 178 | 116 | ||
FS706643.1 | - | 2 - 3 | 195 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |