Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243412.1 | NP_001230341.1 | 4 - 5 | 556 | 96 | |
NM_001243413.1 | NP_001230342.1 | 3 - 4 | 413 | 96 | ||
XM_005655700.3 | XP_005655757.1 | 1196 | 96 | |||
XM_013988430.2 | XP_013843884.1 | 1281 | 96 | |||
XM_013988431.2 | XP_013843885.1 | 455 | 96 | |||
XM_021090919.1 | XP_020946578.1 | 1202 | 96 | |||
XM_021090921.1 | XP_020946580.1 | 445 | 96 | |||
XM_021090922.1 | XP_020946581.1 | 1281 | 96 | |||
XM_021090923.1 | XP_020946582.1 | 270 | 96 | |||
XM_021090924.1 | XP_020946583.1 | 1007 | 96 | |||
GenBank mRNA | AK343836.1 | - | 4 - 5 | 540 | 96 | |
AK400126.1 | - | 4 - 5 | 539 | 96 | ||
FS706643.1 | - | 4 - 5 | 556 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |