Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243421.1 | NP_001230350.1 | 6 - 7 | 544 | 82 | |
NM_001243422.1 | NP_001230351.1 | 6 - 7 | 502 | 82 | ||
XM_021065093.1 | XP_020920752.1 | 697 | 82 | |||
GenBank mRNA | AK351132.1 | - | 6 - 7 | 502 | 82 | |
FD631996.1 | - | 6 - 7 | 544 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |