Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243441.1 | NP_001230370.1 | 3 - 4 | 474 | 68 | |
GenBank mRNA | AK231739.1 | - | 3 - 4 | 474 | 68 | |
AK232111.1 | - | 3 - 4 | 464 | 68 | ||
AK350256.1 | - | 3 - 4 | 442 | 68 | ||
AK352279.1 | - | 3 - 4 | 450 | 68 | ||
AY609894.1 | - | 3 - 4 | 452 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |