Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243444.1 | NP_001230373.1 | 2 - 3 | 393 | 61 | |
XM_021065094.1 | XP_020920753.1 | 626 | 61 | |||
XM_021065095.1 | XP_020920754.1 | 569 | 61 | |||
GenBank mRNA | AK391886.1 | - | 2 - 3 | 413 | 61 | |
AK399963.1 | - | 2 - 3 | 477 | 61 | ||
AY609541.1 | - | 2 - 3 | 456 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |