Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243468.1 | NP_001230397.1 | 1 - 2 | 177 | 77 | |
XM_005663237.2 | XP_005663294.1 | 172 | 77 | |||
GenBank mRNA | AK345954.1 | - | 1 - 2 | 177 | 77 | |
AK345986.1 | - | 1 - 2 | 178 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |