Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243468.1 | NP_001230397.1 | 2 - 3 | 210 | 98 | |
XM_005663237.2 | XP_005663294.1 | 205 | 98 | |||
GenBank mRNA | AK345954.1 | - | 2 - 3 | 210 | 98 | |
AK345986.1 | - | 2 - 3 | 211 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |