Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243472.1 | NP_001230401.1 | 4 - 5 | 891 | 105 | |
XM_005671056.3 | XP_005671113.2 | 1124 | 105 | |||
XM_013983312.2 | XP_013838766.1 | 1054 | 105 | |||
XM_013983314.2 | XP_013838768.1 | 792 | 105 | |||
XM_021071924.1 | XP_020927583.1 | 2861 | 105 | |||
XM_021071925.1 | XP_020927584.1 | 3344 | 105 | |||
XM_021071926.1 | XP_020927585.1 | 1017 | 105 | |||
XM_021071927.1 | XP_020927586.1 | 805 | 105 | |||
GenBank mRNA | AK346500.1 | - | 4 - 5 | 891 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |