Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243487.1 | NP_001230416.1 | 3 - 4 | 221 | 105 | |
GenBank mRNA | AK234247.1 | - | 3 - 4 | 199 | 105 | |
AK234792.1 | - | 3 - 4 | 195 | 105 | ||
AK235981.1 | - | 3 - 4 | 206 | 105 | ||
AK236925.1 | - | 3 - 4 | 198 | 105 | ||
AK390014.1 | - | 3 - 4 | 198 | 105 | ||
AK395190.1 | - | 3 - 4 | 201 | 105 | ||
AK400231.1 | - | 3 - 4 | 216 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |