Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243500.1 | NP_001230429.1 | 3 - 4 | 427 | 99 | |
GenBank mRNA | AK344527.1 | - | 3 - 4 | 427 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |