Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243520.1 | NP_001230449.1 | 1 - 2 | 261 | 68 | |
GenBank mRNA | AK392640.1 | - | 1 - 2 | 101 | 68 | |
FD597318.1 | - | 1 - 2 | 261 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |