Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243527.1 | NP_001230456.1 | 2 - 3 | 404 | 106 | |
GenBank mRNA | AK347144.1 | - | 2 - 3 | 414 | 106 | |
DB803167.1 | - | 2 - 3 | 403 | 106 | ||
DT331792.1 | - | 2 - 3 | 434 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |