Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243528.1 | NP_001230457.1 | 3 - 4 | 711 | 108 | |
XM_021088564.1 | XP_020944223.1 | 1192 | 108 | |||
GenBank mRNA | AK236429.1 | - | 3 - 4 | 447 | 108 | |
AK240283.1 | - | 3 - 4 | 483 | 108 | ||
AK400153.1 | - | 4 - 5 | 930 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |