Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243530.1 | NP_001230459.1 | 3 - 4 | 274 | 92 | |
GenBank mRNA | AK344778.1 | - | 3 - 4 | 275 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |