Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243549.1 | NP_001230478.1 | 4 - 5 | 1139 | 79 | |
XM_013988168.2 | XP_013843622.1 | 1532 | 79 | |||
GenBank mRNA | DN114439.1 | - | 2 - 3 | 492 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |