Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243608.1 | NP_001230537.1 | 5 - 6 | 658 | 71 | |
GenBank mRNA | AK236403.1 | - | 4 - 5 | 473 | 71 | |
FS690453.1 | - | 5 - 6 | 650 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |