Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243618.1 | NP_001230547.1 | 2 - 3 | 626 | 92 | |
GenBank mRNA | AK239456.1 | - | 2 - 3 | 626 | 92 | |
AK350186.1 | - | 3 - 4 | 559 | 92 | ||
AK396939.1 | - | 2 - 3 | 960 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |