Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243627.1 | NP_001230556.1 | 3 - 4 | 525 | 59 | |
GenBank mRNA | AK344213.1 | - | 3 - 4 | 354 | 59 | |
AK348669.1 | - | 3 - 4 | 368 | 59 | ||
AY609785.1 | - | 3 - 4 | 524 | 59 | ||
EW202606.2 | - | 3 - 4 | 521 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |