Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243635.1 | NP_001230564.1 | 2 - 3 | 284 | 102 | |
XM_005655718.3 | XP_005655775.1 | 351 | 102 | |||
GenBank mRNA | AK343312.1 | - | 2 - 3 | 284 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |