Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243663.1 | NP_001230592.1 | 3 - 4 | 245 | 102 | |
XM_021072012.1 | XP_020927671.1 | 1645 | 102 | |||
XM_021072013.1 | XP_020927672.1 | 575 | 102 | |||
GenBank mRNA | AK239039.1 | - | 3 - 4 | 245 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |