Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243668.1 | NP_001230597.1 | 1 - 2 | 38 | 78 | |
GenBank mRNA | AK389884.1 | - | 1 - 2 | 96 | 78 | |
FS665696.1 | - | 1 - 2 | 39 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Sus scrofa Probe spans exons |