Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243691.1 | NP_001230620.1 | 1 - 2 | 76 | 131 | |
GenBank mRNA | AK349998.1 | - | 1 - 2 | 76 | 131 | |
AK392015.1 | - | 1 - 2 | 111 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |