Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243698.1 | NP_001230627.1 | 2 - 3 | 286 | 95 | |
GenBank mRNA | AK240417.1 | - | 2 - 3 | 285 | 95 | |
AK350993.1 | - | 2 - 3 | 290 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |