Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243714.1 | NP_001230643.1 | 3 - 4 | 725 | 87 | |
GenBank mRNA | AK230631.1 | - | 3 - 4 | 725 | 87 | |
AK389835.1 | - | 3 - 4 | 725 | 87 | ||
EV976475.2 | - | 1 - 2 | 143 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |