Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243806.1 | NP_001230735.1 | 2 - 3 | 530 | 74 | |
XM_021066080.1 | XP_020921739.1 | 592 | 74 | |||
XM_021066081.1 | XP_020921740.1 | 592 | 74 | |||
XM_021066082.1 | XP_020921741.1 | 592 | 74 | |||
XM_021066083.1 | XP_020921742.1 | 592 | 74 | |||
GenBank mRNA | AK231645.1 | - | 2 - 3 | 530 | 74 | |
AK240440.1 | - | 2 - 3 | 530 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |