Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243814.1 | NP_001230743.1 | 4 - 5 | 907 | 87 | |
GenBank mRNA | AK349684.1 | - | 4 - 5 | 907 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |