Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243822.1 | NP_001230751.1 | 4 - 5 | 722 | 85 | |
GenBank mRNA | AK345819.1 | - | 4 - 5 | 722 | 85 | |
AK351211.1 | - | 4 - 5 | 740 | 85 | ||
AK389301.1 | - | 4 - 5 | 735 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |