Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRACR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243838.1 | NP_001230767.1 | 5 - 6 | 691 | 80 | |
XM_013989677.2 | XP_013845131.1 | 1369 | 80 | |||
XM_013989679.2 | XP_013845133.1 | 1333 | 80 | |||
XM_013989682.2 | XP_013845136.1 | 1369 | 80 | |||
XM_013989684.2 | XP_013845138.1 | 1461 | 80 | |||
GenBank mRNA | AK350005.1 | - | 5 - 6 | 691 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |