Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243870.1 | NP_001230799.1 | 2 - 3 | 268 | 62 | |
GenBank mRNA | BX924283.2 | - | 2 - 3 | 246 | 62 | |
FD609375.1 | - | 2 - 3 | 268 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |