Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243893.1 | NP_001230822.1 | 2 - 3 | 445 | 87 | |
GenBank mRNA | AK237063.1 | - | 2 - 3 | 445 | 87 | |
AK391421.1 | - | 2 - 3 | 520 | 87 | ||
AK399602.1 | - | 1 - 2 | 109 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |