Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243905.1 | NP_001230834.1 | 1 - 2 | 107 | 88 | |
GenBank mRNA | AK232815.1 | - | 1 - 2 | 107 | 88 | |
AY609754.1 | - | 1 - 2 | 106 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |