Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243950.1 | NP_001230879.1 | 4 - 5 | 627 | 90 | |
XM_021074608.1 | XP_020930267.1 | 747 | 90 | |||
XM_021074609.1 | XP_020930268.1 | 1297 | 90 | |||
XM_021074610.1 | XP_020930269.1 | 939 | 90 | |||
XM_021074612.1 | XP_020930271.1 | 641 | 90 | |||
XM_021074613.1 | XP_020930272.1 | 703 | 90 | |||
XM_021074614.1 | XP_020930273.1 | 1957 | 90 | |||
GenBank mRNA | AK232101.1 | - | 4 - 5 | 627 | 90 | |
AK232384.1 | - | 4 - 5 | 518 | 90 | ||
AK232856.1 | - | 5 - 6 | 694 | 90 | ||
AK396251.1 | - | 4 - 5 | 532 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |