Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244060.1 | NP_001230989.1 | 4 - 5 | 582 | 82 | |
GenBank mRNA | AK234001.1 | - | 4 - 5 | 551 | 82 | |
AY609814.1 | - | 4 - 5 | 540 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |