Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244096.1 | NP_001231025.1 | 1 - 2 | 117 | 65 | |
XM_021079411.1 | XP_020935070.1 | 189 | 65 | |||
GenBank mRNA | AK235135.1 | - | 1 - 2 | 117 | 65 | |
AK236451.1 | - | 1 - 2 | 120 | 65 | ||
AK236585.1 | - | 1 - 2 | 122 | 65 | ||
AK394814.1 | - | 1 - 2 | 117 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |