Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244099.1 | NP_001231028.1 | 2 - 3 | 126 | 63 | |
XM_005662155.3 | XP_005662212.1 | 246 | 63 | |||
XM_021085649.1 | XP_020941308.1 | 472 | 63 | |||
XM_021085650.1 | XP_020941309.1 | 474 | 63 | |||
XM_021085651.1 | XP_020941310.1 | 374 | 63 | |||
XM_021085652.1 | XP_020941311.1 | 343 | 63 | |||
XM_021085653.1 | XP_020941312.1 | 344 | 63 | |||
GenBank mRNA | BE030992.1 | - | 2 - 3 | 126 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |