Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244099.1 | NP_001231028.1 | 3 - 4 | 234 | 77 | |
XM_005662155.3 | XP_005662212.1 | 354 | 77 | |||
XM_021085649.1 | XP_020941308.1 | 580 | 77 | |||
XM_021085650.1 | XP_020941309.1 | 582 | 77 | |||
XM_021085651.1 | XP_020941310.1 | 482 | 77 | |||
XM_021085652.1 | XP_020941311.1 | 451 | 77 | |||
XM_021085653.1 | XP_020941312.1 | 452 | 77 | |||
GenBank mRNA | BE030992.1 | - | 3 - 4 | 234 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |