Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244104.1 | NP_001231033.1 | 3 - 4 | 611 | 59 | |
GenBank mRNA | AK234563.1 | - | 3 - 4 | 610 | 59 | |
AK235641.1 | - | 3 - 4 | 277 | 59 | ||
BW970534.1 | - | 3 - 4 | 264 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |