Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244146.2 | NP_001231075.1 | 3 - 4 | 428 | 70 | |
XM_005653601.2 | XP_005653658.1 | 1283 | 70 | |||
XM_005653603.3 | XP_005653660.1 | 694 | 70 | |||
XM_005653605.2 | XP_005653662.1 | 1161 | 70 | |||
XM_005653610.3 | XP_005653667.1 | 910 | 70 | |||
XM_005653613.3 | XP_005653670.1 | 695 | 70 | |||
XM_013998341.1 | XP_013853795.1 | 1059 | 70 | |||
XM_013998376.2 | XP_013853830.1 | 1285 | 70 | |||
XM_013998382.2 | XP_013853836.1 | 1285 | 70 | |||
XM_021100333.1 | XP_020955992.1 | 2199 | 70 | |||
XM_021100334.1 | XP_020955993.1 | 1093 | 70 | |||
XM_021100335.1 | XP_020955994.1 | 1371 | 70 | |||
XM_021100336.1 | XP_020955995.1 | 918 | 70 | |||
XM_021100337.1 | XP_020955996.1 | 925 | 70 | |||
XM_021100338.1 | XP_020955997.1 | 1019 | 70 | |||
XM_021100339.1 | XP_020955998.1 | 869 | 70 | |||
XM_021100340.1 | XP_020955999.1 | 833 | 70 | |||
XM_021100341.1 | XP_020956000.1 | 751 | 70 | |||
XM_021100342.1 | XP_020956001.1 | 562 | 70 | |||
XM_021100343.1 | XP_020956002.1 | 1601 | 70 | |||
GenBank mRNA | AK238426.1 | - | 3 - 4 | 429 | 70 | |
AK238452.1 | - | 3 - 4 | 352 | 70 | ||
AK391331.1 | - | 5 - 6 | 665 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |