Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244147.1 | NP_001231076.1 | 2 - 3 | 352 | 84 | |
GenBank mRNA | AK231884.1 | - | 2 - 3 | 354 | 84 | |
AK232862.1 | - | 2 - 3 | 352 | 84 | ||
AK345822.1 | - | 2 - 3 | 318 | 84 | ||
AK350259.1 | - | 2 - 3 | 317 | 84 | ||
AY610158.1 | - | 2 - 3 | 326 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |